Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-(o-Tolyl)thiourea, 97+%

Click to view available options
Quantity:
10g
50g
250g
614-78-8
C8H10N2S
166.242
MFCD00041162
ACLZYRNSDLQOIA-UHFFFAOYSA-N
o-tolylthiourea,2-methylphenyl thiourea,n-o-tolyl thiourea,1-2-methylphenyl thiourea,n-2-methylphenyl thiourea,1-2-methylphenyl-2-thiourea,2-tolylthiourea,1-o-tolyl-2-thiourea,thiourea, 2-methylphenyl,1-o-tolyl thiourea
2787703
(2-methylphenyl)thiourea
CC1=CC=CC=C1NC(=S)N
614-78-8
C8H10N2S
166.242
MFCD00041162
ACLZYRNSDLQOIA-UHFFFAOYSA-N
o-tolylthiourea,2-methylphenyl thiourea,n-o-tolyl thiourea,1-2-methylphenyl thiourea,n-2-methylphenyl thiourea,1-2-methylphenyl-2-thiourea,2-tolylthiourea,1-o-tolyl-2-thiourea,thiourea, 2-methylphenyl,1-o-tolyl thiourea
2787703
(2-methylphenyl)thiourea
CC1=CC=CC=C1NC(=S)N
Quantity 10g
Odor Odorless
Melting Point 155°C to 158°C
UN Number UN2811
Beilstein 2086802
Formula Weight 166.25
Percent Purity ≥97%
Chemical Name or Material N-(o-Tolyl)thiourea

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P280f-P301+P310-P330-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : YU2975000

TSCA : TSCA