Alfa Aesar™ n-Octacosane, 99%

Manufacturer: Alfa Aesar™ 4246009

Quantity 10g

 View more versions of this product

Catalog No. AA4246009

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 630-02-4
    Formula Weight 394.77
    MDL Number MFCD00009355
    Chemical Name or Material n-Octacosane

    Specifications

    Formula Weight 394.77
    Quantity 10g
    Flash Point 227°C (441°F)
    MDL Number MFCD00009355
    Melting Point 61° to 63°C
    TSCA TSCA
    Boiling Point 440°C
    Physical Form Crystalline
    CAS 630-02-4
    Chemical Name or Material n-Octacosane
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit