Alfa Aesar™ n-Octadecanophenone, 99%

Manufacturer: Alfa Aesar™ H53394-06

 View more versions of this product

Catalog No. AAH5339406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6786-36-3
    Molecular Formula C24H40O
    Formula Weight 344.58g/mol
    MDL Number MFCD00010700
    Chemical Name or Material n-Octadecanophenone

    Specifications

    Melting Point 65° to 67°C
    Boiling Point 425°C
    Quantity 5g
    CAS 6786-36-3
    Molecular Formula C24H40O
    Formula Weight 344.58g/mol
    MDL Number MFCD00010700
    Chemical Name or Material n-Octadecanophenone
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit