Alfa Aesar™ n-Octadecenylsuccinic anhydride

Manufacturer: Alfa Aesar™ 3195718

Quantity 50g

 View more versions of this product

Catalog No. AA3195718

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 28777-98-2
    Formula Weight 350.52
    Chemical Name or Material n-Octadecenylsuccinic anhydride

    Specifications

    Formula Weight 350.52
    Health Hazard 3 GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.
    Melting Point 64°C
    Boiling Point 245° to 255°C (5mmHg)
    CAS 28777-98-2
    Health Hazard 1 Warning
    TSCA TSCA
    Physical Form Crystalline
    Quantity 50g
    Health Hazard 2 GHS H Statement
    H319
    Causes serious eye irritation.
    Chemical Name or Material n-Octadecenylsuccinic anhydride

    GHS H Statement
    H319
    Causes serious eye irritation.

    GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit