Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octadecyl sulfate, sodium salt, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
1120-04-3
C18H37NaO4S
372.55
MFCD00007469
NWZBFJYXRGSRGD-UHFFFAOYSA-M
sodium stearyl sulfate,octadecyl sulfate sodium salt,sodium octadecyl sulfate,octadecyl sodium sulfate,sulfuric acid, monooctadecyl ester, sodium salt,sodium octadecyl sulphate,sodium monostearyl sulfate,unii-t4eww0y4ej,sodium n-octadecyl sulfate,sodium monooctadecyl sulfate
23673604
sodium;octadecyl sulfate
CCCCCCCCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]
1120-04-3
C18H37NaO4S
372.55
MFCD00007469
NWZBFJYXRGSRGD-UHFFFAOYSA-M
sodium stearyl sulfate,octadecyl sulfate sodium salt,sodium octadecyl sulfate,octadecyl sodium sulfate,sulfuric acid, monooctadecyl ester, sodium salt,sodium octadecyl sulphate,sodium monostearyl sulfate,unii-t4eww0y4ej,sodium n-octadecyl sulfate,sodium monooctadecyl sulfate
23673604
sodium;octadecyl sulfate
CCCCCCCCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]
Quantity 1g
Color Undesignated
Packaging Glass bottle
Melting Point 211.0°C to 213.0°C
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (Ex Sulfated Ash)
Linear Formula CH3(CH2)17OSO3Na
Solubility Information Solubility in water: soluble.
Formula Weight 372.55
Physical Form Powder
Percent Purity 98%
Chemical Name or Material n-Octadecyl sulfate, sodium salt
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 214-295-