Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octane, 99+%, Acros Organics™

Click to view available options
Quantity:
2mL
2.5L
25L
Packaging:
Glass Bottle
Metal drum
111-65-9
C8H18
114.23
MFCD00009556
TVMXDCGIABBOFY-UHFFFAOYSA-N
n-octane,oktan,oktanen,ottani,oktanen dutch,heptane, methyl,oktan polish,ottani italian,octane, all isomers,unii-x1rv0b2fjv
356
CHEBI:17590
octane
CCCCCCCC
111-65-9
C8H18
114.23
MFCD00009556
TVMXDCGIABBOFY-UHFFFAOYSA-N
n-octane,oktan,oktanen,ottani,oktanen dutch,heptane, methyl,oktan polish,ottani italian,octane, all isomers,unii-x1rv0b2fjv
356
CHEBI:17590
octane
CCCCCCCC
Viscosity 0.55 mPa.s (20°C)
Flash Point 13°C
Specific Gravity 0.708
Packaging Metal drum
Infrared Spectrum Authentic
Refractive Index 1.3965 to 1.3985
Color Colorless
Vapor Pressure 14mbar at 20°C
Melting Point -57.0°C
Quantity 25L
Residue after Evaporation 0.05% max.
Density 0.7080g/mL
Boiling Point 125.0°C to 127.0°C
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula CH3(CH2)6CH3
Beilstein 01, 159
Merck Index 15, 6838
Solubility Information Solubility in water: 0.0007g/L (20°C). Other solubilities: soluble in alcohol and acetone, freely miscible with ether, benzene, petroleum, ether and gasoline
Formula Weight 114.23
Physical Form Liquid
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material n-Octane
Show More Show Less

GHS H Statement:
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger