Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Octanoyl-D-erythro-sphingosine, ≥99%, synthetical, Acros Organics

Click to view available options
Quantity:
50mg
Packaging:
Glass bottle
74713-59-0
C26H51NO3
425.698
MFCD00236496
APDLCSPGWPLYEQ-WRBRXSDHSA-N
c8 ceramide,n-octanoylsphingosine,c-8 ceramide,d-erythro-sphingosine, n-octanoyl,n-2s,3r,4e-1,3-dihydroxyoctadec-4-en-2-yl octanamide,c8-ceramide,n-octanoyl-d-erythro-sphingosine,n-octanoyl sphingosine,n-octanoylsphing-4-enine,n-octanoyl-sphing-4-enine
5702614
CHEBI:45815
N-[(E,2S,3R)-1,3-dihydroxyoctadec-4-en-2-yl]octanamide
CCCCCCCCCCCCCC=CC(C(CO)NC(=O)CCCCCCC)O
74713-59-0
C26H51NO3
425.698
MFCD00236496
APDLCSPGWPLYEQ-WRBRXSDHSA-N
c8 ceramide,n-octanoylsphingosine,c-8 ceramide,d-erythro-sphingosine, n-octanoyl,n-2s,3r,4e-1,3-dihydroxyoctadec-4-en-2-yl octanamide,c8-ceramide,n-octanoyl-d-erythro-sphingosine,n-octanoyl sphingosine,n-octanoylsphing-4-enine,n-octanoyl-sphing-4-enine
5702614
CHEBI:45815
N-[(E,2S,3R)-1,3-dihydroxyoctadec-4-en-2-yl]octanamide
CCCCCCCCCCCCCC=CC(C(CO)NC(=O)CCCCCCC)O
Packaging Glass bottle
Infrared Spectrum Authentic
Color White
Quantity 50mg
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Formula Weight 425.69
Physical Form Powder
Percent Purity ≥99.0%
Chemical Name or Material N-Octanoyl-D-erythro-sphingosine, 99+%
Show More Show Less