Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ n-Octyl-beta-D-thioglucopyranoside, Electrophoresis Grade, ≥98%

Click to view available options
Quantity:
2.5g
5g
10g
85618-21-9
C14H28O5S
308.433
MFCD00012189
CGVLVOOFCGWBCS-DGTMBMJNSA-N
octyl thioglucopyranoside,octyl-i 2-d-1-thioglucopyranoside
53720324
(2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-octylsulfanyloxane-3,4,5-triol
CCCCCCCCSC1C(C(C(C(O1)CO)O)O)O
85618-21-9
C14H28O5S
308.433
MFCD00012189
CGVLVOOFCGWBCS-DGTMBMJNSA-N
octyl thioglucopyranoside,octyl-i 2-d-1-thioglucopyranoside
53720324
(2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-octylsulfanyloxane-3,4,5-triol
CCCCCCCCSC1C(C(C(C(O1)CO)O)O)O
Color White
Quantity 5g
Melting Point 128°C
Beilstein 4182783
Formula Weight 308.44
Physical Form Crystalline Powder
Percent Purity ≥98%
Grade Electrophoresis
Chemical Name or Material n-Octyl-beta-D-thioglucopyranoside