Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ n-Octyl butyrate, 97%

Click to view available options
Quantity:
50g
250g
1000g
110-39-4
C12H24O2
200.322
MFCD00048939
PWLNAUNEAKQYLH-UHFFFAOYSA-N
octyl butyrate,butanoic acid, octyl ester,n-octyl butyrate,butyric acid, octyl ester,n-octyl n-butyrate,n-octyl butanoate,unii-5yeu4o369l,butyric acid, octyl ester 8ci,octyl butyrate natural,fema no. 2807
61030
octyl butanoate
CCCCCCCCOC(=O)CCC
110-39-4
C12H24O2
200.322
MFCD00048939
PWLNAUNEAKQYLH-UHFFFAOYSA-N
octyl butyrate,butanoic acid, octyl ester,n-octyl butyrate,butyric acid, octyl ester,n-octyl n-butyrate,n-octyl butanoate,unii-5yeu4o369l,butyric acid, octyl ester 8ci,octyl butyrate natural,fema no. 2807
61030
octyl butanoate
CCCCCCCCOC(=O)CCC
Quantity 1000g
Formula Weight 172.27
Percent Purity 97%
Chemical Name or Material n-Octyl butyrate