Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ n-Octyl butyrate, 97%
Click to view available options
Quantity:
50g
250g
1000g
Specifications
Quantity | 250g |
Formula Weight | 172.27 |
Percent Purity | 97% |
Chemical Name or Material | n-Octyl butyrate |
Safety and Handling
EINECSNumber : 203-762-4
TSCA : Yes
Recommended Storage : Ambient temperatures