Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ n-Octyl-β-D-glucopyranoside, ULTROL™ Grade, Calbiochem™,

Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass Bottle
29836-26-8
C14H28O6
292.372
HEGSGKPQLMEBJL-RKQHYHRCSA-N
OG, n-Octylglucoside
62852
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-octoxyoxane-3,4,5-triol
CCCCCCCCOC1C(C(C(C(O1)CO)O)O)O
29836-26-8
C14H28O6
292.372
HEGSGKPQLMEBJL-RKQHYHRCSA-N
OG, n-Octylglucoside
62852
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-octoxyoxane-3,4,5-triol
CCCCCCCCOC1C(C(C(C(O1)CO)O)O)O
Packaging Glass Bottle
Percent Purity ≥99% by TLC
Grade ULTROL
Color White
Quantity 250mg