Alfa Aesar™ n-Octyl L-2-pyrrolidinone-5-carboxylate, 98%

Manufacturer: Alfa Aesar™ B2183814

Quantity 25g

 View more versions of this product

Catalog No. AAB2183814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4931-70-8
    Molecular Formula C13H23NO3
    Formula Weight 241.33
    MDL Number MFCD00236053
    Beilstein 478315
    Synonym n-Octyl L-pyroglutamate; L-Pyroglutamic acid n-octyl ester
    Chemical Name or Material n-Octyl L-2-pyrrolidinone-5-carboxylate

    Specifications

    Formula Weight 241.33
    Boiling Point 140°C (0.06mmHg)
    Quantity 25g
    Molecular Formula C13H23NO3
    MDL Number MFCD00236053
    CAS 4931-70-8
    Beilstein 478315
    Synonym n-Octyl L-pyroglutamate; L-Pyroglutamic acid n-octyl ester
    Chemical Name or Material n-Octyl L-2-pyrrolidinone-5-carboxylate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit