Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylene glycol monooctyl ether, 98%, Acros Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass Bottle
19327-39-0
C16H34O5
306.443
FEOZZFHAVXYAMB-UHFFFAOYSA-N
3,6,9,12-tetraoxaeicosan-1-ol,3,6,9,12-tetraoxaicosan-1-ol,tetraethylene glycol monooctyl ether,hydroxyethyloxy tri ethyloxy octane,tetraethylene glycol monoctyl ether,c8e,octyltetraglycol,octyltetraethyleneoxide,d06pvf,octyl tetraethylene glycol ether
5414
CHEBI:41323
2-[2-[2-(2-octoxyethoxy)ethoxy]ethoxy]ethanol
CCCCCCCCOCCOCCOCCOCCO
19327-39-0
C16H34O5
306.443
FEOZZFHAVXYAMB-UHFFFAOYSA-N
3,6,9,12-tetraoxaeicosan-1-ol,3,6,9,12-tetraoxaicosan-1-ol,tetraethylene glycol monooctyl ether,hydroxyethyloxy tri ethyloxy octane,tetraethylene glycol monoctyl ether,c8e,octyltetraglycol,octyltetraethyleneoxide,d06pvf,octyl tetraethylene glycol ether
5414
CHEBI:41323
2-[2-[2-(2-octoxyethoxy)ethoxy]ethoxy]ethanol
CCCCCCCCOCCOCCOCCOCCO
Packaging Glass Bottle
Boiling Point 170.0°C (0.4 mmHg)
Flash Point 179°C
Quantity 1g
Density 0.9600g/mL
Specific Gravity 0.96
Refractive Index 1.4464 to 1.4484
Melting Point -2.0°C
Assay Percent Range 94%
Linear Formula CH3(CH2)7OCH2CH2OCH2CH2OCH2CH2OCH2CH2OH
Solubility Information Solubility in water: soluble.
Formula Weight 306.39
Percent Purity 98%
Chemical Name or Material Tetraethylene glycol monooctyl ether, 94%
Show More Show Less