Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octylamine, 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
Plastic Jerrycan
111-86-4
C8H19N
129.24
IOQPZZOEVPZRBK-UHFFFAOYSA-N
octylamine,n-octylamine,1-aminooctane,1-octanamine,1-octylamine,caprylamine,caprylylamine,armeen 8,n-octylamine, mono,armeen 8d
8143
CHEBI:7728
octan-1-amine
CCCCCCCCN
111-86-4
C8H19N
129.24
IOQPZZOEVPZRBK-UHFFFAOYSA-N
octylamine,n-octylamine,1-aminooctane,1-octanamine,1-octylamine,caprylamine,caprylylamine,armeen 8,n-octylamine, mono,armeen 8d
8143
CHEBI:7728
octan-1-amine
CCCCCCCCN
Viscosity 1.39 mPa.s (20°C)
Specific Gravity 0.782
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Boiling Point 179.0°C
Water 0.2% max.
Flash Point 60°C
Density 0.7820g/mL
Refractive Index 1.4280 to 1.4300
Melting Point -1.0°C
Color Undesignated
Assay Percent Range 99+%
Linear Formula CH3(CH2)7NH2
Beilstein 04,196
Solubility Information Solubility in water: 0.2g/L water (25°C). Other solubilities: soluble in most common organic solvents
Formula Weight 129.24
Physical Form Liquid
Percent Purity ≥99%
Chemical Name or Material n-Octylamine
Show More Show Less