Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octylamine, 99+%, Acros Organics™

Click to view available options
Quantity:
2kg
111-86-4
C8H19N
129.24
IOQPZZOEVPZRBK-UHFFFAOYSA-N
octylamine,n-octylamine,1-aminooctane,1-octanamine,1-octylamine,caprylamine,caprylylamine,armeen 8,n-octylamine, mono,armeen 8d
8143
CHEBI:7728
octan-1-amine
CCCCCCCCN
111-86-4
C8H19N
129.24
IOQPZZOEVPZRBK-UHFFFAOYSA-N
octylamine,n-octylamine,1-aminooctane,1-octanamine,1-octylamine,caprylamine,caprylylamine,armeen 8,n-octylamine, mono,armeen 8d
8143
CHEBI:7728
octan-1-amine
CCCCCCCCN
Viscosity 1.39 mPa.s (20°C)
Quantity 2kg
Melting Point -1.0°C
Boiling Point 179.0°C
Infrared Spectrum Authentic
Specific Gravity 0.782
Water 0.2% max.
Flash Point 60°C
Refractive Index 1.4280 to 1.4300
Density 0.7820g/mL
Assay Percent Range 99% min. (GC)
Linear Formula CH3(CH2)7NH2
Beilstein 04,196
Solubility Information Solubility in water: 0.2g/L water (25°C). Other solubilities: soluble in most common organic solvents
Formula Weight 129.24
Percent Purity 99+%
Chemical Name or Material n-Octylamine
Show More Show Less

GHS H Statement
Flammable liquid and vapour.
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
May cause respiratory irritation.
Very toxic to aquatic l

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Danger

EINECSNumber : 203-916-