Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octyltrichlorosilane, 97%, Thermo Scientific Chemicals

Catalog Number AAB2375336
Click to view available options
Quantity:
25 g
100 g
500 g
5283-66-9
C8H17Cl3Si
247.659
MFCD00000488
RCHUVCPBWWSUMC-UHFFFAOYSA-N
trichloro octyl silane, n-octyltrichlorosilane, octyltrichlorosilane, silane, trichlorooctyl, trichloro-n-octylsilane, unii-jv18i1wcu8, jv18i1wcu8, silane, octyltrichloro, octyltrichlorosilane un1801 corrosive
21354
trichloro(octyl)silane
CCCCCCCC[Si](Cl)(Cl)Cl
5283-66-9
C8H17Cl3Si
247.659
MFCD00000488
RCHUVCPBWWSUMC-UHFFFAOYSA-N
trichloro octyl silane, n-octyltrichlorosilane, octyltrichlorosilane, silane, trichlorooctyl, trichloro-n-octylsilane, unii-jv18i1wcu8, jv18i1wcu8, silane, octyltrichloro, octyltrichlorosilane un1801 corrosive
21354
trichloro(octyl)silane
CCCCCCCC[Si](Cl)(Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 225°C to 227°C
Quantity 500 g
UN Number UN1801
Beilstein 1744230
Flash Point 96°C (204°F)
Formula Weight 247.67
Percent Purity 97%
Odor Irritating
Refractive Index 1.447
Sensitivity Moisture sensitive
Density 1.073
Chemical Name or Material n-Octyltrichlorosilane
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: OCTYLTRICHLOROSILANE
EINECSNumber 226-112-1
RTECSNumber VV4700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit