Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-(p-Toluenesulfonyl)-L-glutamic acid, 97%

Click to view available options
Quantity:
25g
100g
500g
4816-80-2
C12H15NO6S
301.313
MFCD00020545
KKOZKXBAPIYWAT-JTQLQIEISA-N
s-2-4-methylphenylsulfonamido pentanedioic acid,tosyl-l-glutamic acid,n-tosyl-l-glutamic acid,n-p-tolylsulphonyl-l-glutamic acid,+-n-tosyl-l-glutamic acid,2s-2-4-methylbenzenesulfonamido pentanedioic acid,l-glutamic acid, n-4-methylphenyl sulfonyl,tos-glu-oh,2-4-methylphenylsulfonamido pentanedioic acid,ksc493e0f
2063452
(2S)-2-[(4-methylphenyl)sulfonylamino]pentanedioic acid
CC1=CC=C(C=C1)S(=O)(=O)NC(CCC(=O)O)C(=O)O
4816-80-2
C12H15NO6S
301.313
MFCD00020545
KKOZKXBAPIYWAT-JTQLQIEISA-N
s-2-4-methylphenylsulfonamido pentanedioic acid,tosyl-l-glutamic acid,n-tosyl-l-glutamic acid,n-p-tolylsulphonyl-l-glutamic acid,+-n-tosyl-l-glutamic acid,2s-2-4-methylbenzenesulfonamido pentanedioic acid,l-glutamic acid, n-4-methylphenyl sulfonyl,tos-glu-oh,2-4-methylphenylsulfonamido pentanedioic acid,ksc493e0f
2063452
(2S)-2-[(4-methylphenyl)sulfonylamino]pentanedioic acid
CC1=CC=C(C=C1)S(=O)(=O)NC(CCC(=O)O)C(=O)O
Beilstein 2223843
Formula Weight 301.31
Physical Form Solid
Percent Purity 97%
Density 1.427g/mL
Melting Point 130°C to 132°C
Quantity 100g
Chemical Name or Material N-(p-Toluenesulfonyl)-L-glutamic acid