Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-(p-Toluenesulfonyl)-L-phenylalanine chloromethyl ketone

Click to view available options
Quantity:
5g
402-71-1
C17H18ClNO3S
351.845
MFCD00000935
MQUQNUAYKLCRME-INIZCTEOSA-N
tpck,tos-phe-ch2cl,tosylphenylalanyl chloromethyl ketone,n-tosyl-l-phenylalanyl chloromethyl ketone,n-tosyl-l-phenylalanine chloromethyl ketone,n-p-tosyl-l-phenylalanine chloromethyl ketone,l-1-tosylamido-2-phenylethyl chloromethyl ketone,l-n-alpha-chloroacetyl phenethyl-p-toluenesulfonamide,tos-phe-chloromethylketone,chembl60718
439647
CHEBI:9642
N-[(2S)-4-chloro-3-oxo-1-phenylbutan-2-yl]-4-methylbenzenesulfonamide
CC1=CC=C(C=C1)S(=O)(=O)NC(CC2=CC=CC=C2)C(=O)CCl
402-71-1
C17H18ClNO3S
351.845
MFCD00000935
MQUQNUAYKLCRME-INIZCTEOSA-N
tpck,tos-phe-ch2cl,tosylphenylalanyl chloromethyl ketone,n-tosyl-l-phenylalanyl chloromethyl ketone,n-tosyl-l-phenylalanine chloromethyl ketone,n-p-tosyl-l-phenylalanine chloromethyl ketone,l-1-tosylamido-2-phenylethyl chloromethyl ketone,l-n-alpha-chloroacetyl phenethyl-p-toluenesulfonamide,tos-phe-chloromethylketone,chembl60718
439647
CHEBI:9642
N-[(2S)-4-chloro-3-oxo-1-phenylbutan-2-yl]-4-methylbenzenesulfonamide
CC1=CC=C(C=C1)S(=O)(=O)NC(CC2=CC=CC=C2)C(=O)CCl

Enzyme inhibitor that inactivates chymotrypsin but not trypsin

Beilstein 2895215
Solubility Information Soluble in DMSO at 10mg/ml
Formula Weight 351.85
Physical Form Powder
Color White
Quantity 5g
Chemical Name or Material N-(p-Toluenesulfonyl)-L-phenylalanine chloromethyl ketone

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : XT5613500

TSCA : TSCA

Recommended Storage : −30° to −10°C