Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Palmitoyl-D-erythro-sphingosine, synthetical, 98% (99+% e.e.), ACROS Organics™

Click to view available options
Quantity:
5mg
25mg
250mg
Packaging:
Glass bottle
24696-26-2
C34H67NO3
537.91
MFCD00056205
YDNKGFDKKRUKPY-LQJZCPKCSA-N
c16-ceramide,d-erythro-c16-ceramide,n-palmitoyl 4-sphingenine,n-palmitoyl-d-sphingosine,d-erythro-4-ceramide,n-palmitoylsphingosinee,d-erythro-n-palmitoylsphingosine,n-hexadecanoyl-d-erythro-ceramide,d-erythro-n-hexadecanoylsphingenine,2s,3r,4e-2-n-palmitoyloctadecasphinga-4-ene
71314645
N-[(2S,3S)-1,3-dihydroxyoctadec-4-en-2-yl]hexadecanamide
CCCCCCCCCCCCCCCC(=O)NC(CO)C(C=CCCCCCCCCCCCCC)O
24696-26-2
C34H67NO3
537.91
MFCD00056205
YDNKGFDKKRUKPY-LQJZCPKCSA-N
c16-ceramide,d-erythro-c16-ceramide,n-palmitoyl 4-sphingenine,n-palmitoyl-d-sphingosine,d-erythro-4-ceramide,n-palmitoylsphingosinee,d-erythro-n-palmitoylsphingosine,n-hexadecanoyl-d-erythro-ceramide,d-erythro-n-hexadecanoylsphingenine,2s,3r,4e-2-n-palmitoyloctadecasphinga-4-ene
71314645
N-[(2S,3S)-1,3-dihydroxyoctadec-4-en-2-yl]hexadecanamide
CCCCCCCCCCCCCCCC(=O)NC(CO)C(C=CCCCCCCCCCCCCC)O
Quantity 25mg
Packaging Glass bottle
Enantiomeric Excess 99.0% min.
Color White
Infrared Spectrum Authentic
Melting Point 94.0°C to 95.0°C
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in chloroform,soluble in dichloromethane,insoluble in acetone
Formula Weight 537.91
Physical Form Crystalline Powder
Percent Purity 98% (99+% E.E.)
Chemical Name or Material N-Palmitoyl-D-erythro-sphingosine, Synthetical, ≥99% ee
Show More Show Less