Alfa Aesar™ n-Pentacosane, 99%

Manufacturer: Alfa Aesar™ 3088014

Quantity 25g

 View more versions of this product

Catalog No. AA3088014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 629-99-2
    Formula Weight 352.67
    MDL Number MFCD00009353
    Chemical Name or Material n-Pentacosane

    Specifications

    Formula Weight 352.67
    Refractive Index 1.4380
    Melting Point 53.3°C
    TSCA TSCA
    Physical Form Wax
    MDL Number MFCD00009353
    CAS 629-99-2
    Density 0.7785g/mL
    Boiling Point 259°C (15mmHg)
    Quantity 25g
    Chemical Name or Material n-Pentacosane
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit