Alfa Aesar™ n-Pentacosane, 99%

Manufacturer: Alfa Aesar™ 3088003

Quantity 1g

 View more versions of this product

Catalog No. AA3088003

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 629-99-2
    Formula Weight 352.67
    MDL Number MFCD00009353
    Chemical Name or Material n-Pentacosane

    Specifications

    Formula Weight 352.67
    Refractive Index 1.4380
    MDL Number MFCD00009353
    CAS 629-99-2
    Density 0.7785g/mL
    Boiling Point 259°C (15mmHg)
    Melting Point 53.3°C
    TSCA TSCA
    Physical Form Wax
    Quantity 1g
    Chemical Name or Material n-Pentacosane
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit