Alfa Aesar™ n-Pentadecane, 99%

Manufacturer: Alfa Aesar™ A1033636

Quantity 500g

 View more versions of this product

Catalog No. AAA1033636

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 629-62-9
    Molecular Formula CH3(CH2)13CH3
    Formula Weight 212.42
    MDL Number MFCD00008990
    Beilstein 1698194
    Chemical Name or Material n-Pentadecane

    Specifications

    Formula Weight 212.42
    Health Hazard 3 GHS P Statement
    P301+P310-P331-P405-P501a
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Beilstein 1698194
    Health Hazard 1 Danger
    Boiling Point 269° to 270°C
    Quantity 500g
    Molecular Formula CH3(CH2)13CH3
    CAS 629-62-9
    MDL Number MFCD00008990
    Health Hazard 2 GHS H Statement
    H304
    May be fatal if swallowed and enters airways.
    RTECS Number RZ1800000
    Flash Point 132°C (269°F)
    Melting Point 9° to 10°C
    Chemical Name or Material n-Pentadecane
    Assay 99%

    Description

    Applications
    n-pentadecane is used as a reference substance for gas chromatography. It is also used as standard materials for the determination of combustion quality of diesel. It is involved in the organic synthesis.

    Solubility
    Immiscible with water.

    Notes
    Incompatible with strong oxidizing agents.

    GHS H Statement
    H304
    May be fatal if swallowed and enters airways.

    GHS P Statement
    P301+P310-P331-P405-P501a
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit