Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Pentane, 95%, technical grade, Thermo Scientific Chemicals

Catalog Number p-4229435
Click to view available options
Quantity:
5 L
25 L
Packaging:
Metal Drum
109-66-0
C5H12
72.15
MFCD00009498
OFBQJSOFQDEBGM-UHFFFAOYSA-N
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
8003
CHEBI:37830
pentane
CCCCC
109-66-0
C5H12
72.15
MFCD00009498
OFBQJSOFQDEBGM-UHFFFAOYSA-N
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
8003
CHEBI:37830
pentane
CCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -130.0°C
Color Colorless
CAS Min % 94.0
CAS Max % 100.0
Density 0.6260g/mL
Boiling Point 36.0°C
Quantity 5 L
Flash Point −49°C
Infrared Spectrum Authentic
Packaging Metal Drum
Physical Form Liquid
Refractive Index 1.358
Assay Percent Range 95%
Specific Gravity 0.626
Linear Formula CH3(CH2)3CH3
Viscosity 0.25 mPa.s (20°C)
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and most other common organic, solvents
Formula Weight 72.15
Percent Purity 95%
Chemical Name or Material n-Pentane
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit