Alfa Aesar™ n-Pentane, 98%

Manufacturer: Alfa Aesar™ 17381K2

Quantity 1L

 View more versions of this product

Catalog No. AA17381K2

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 109-66-0
    Molecular Formula C5H12
    Linear Formula CH3(CH2)3CH3
    Formula Weight 72.15
    MDL Number MFCD00009498
    UN Number UN1265
    Merck Index 14,7116
    Chemical Name or Material n-Pentane

    Specifications

    Formula Weight 72.15
    Assay Percent Range 98%
    Molecular Formula C5H12
    Health Hazard 3 GHS P Statement
    P210-P261-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Nearly odorless
    Flash Point −49° (−57°F)
    Boiling Point 36°C
    Melting Point −130°C
    Health Hazard 2 GHS H Statement
    H225-H304-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    May cause drowsiness or dizziness.
    Density 0.626g/mL
    RTECS Number RZ9450000
    Refractive Index 1.3580
    Physical Form Liquid
    Color Colorless
    Linear Formula CH3(CH2)3CH3
    MDL Number MFCD00009498
    CAS 109-66-0
    UN Number UN1265
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PENTANES
    TSCA TSCA
    Quantity 1L
    Merck Index 14,7116
    Health Hazard 1 Danger
    Chemical Name or Material n-Pentane
    Assay 98%

    GHS H Statement
    H225-H304-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P261-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit