Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Pentane, 99+%, Thermo Scientific™

Click to view available options
Quantity:
100mL
1L
109-66-0
C5H12
72.15
MFCD00009498
OFBQJSOFQDEBGM-UHFFFAOYSA-N
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
8003
CHEBI:37830
pentane
CCCCC
109-66-0
C5H12
72.15
MFCD00009498
OFBQJSOFQDEBGM-UHFFFAOYSA-N
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
8003
CHEBI:37830
pentane
CCCCC
Linear Formula CH3(CH2)3CH3
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and most other common organic, solvents
Formula Weight 72.15
Percent Purity ≥99%
Grade Extra Dry over Molecular Sieve
Density 0.6260g/mL
Refractive Index 1.358
Solubility Solubility in water: insoluble. Other solubilities: soluble in alcohol and most other common organic, solvents
Specific Gravity 0.626
Water 0.005% max.
Flash Point −49°C
Viscosity 0.25 mPa.s (20°C)
Boiling Point 36.0°C
Melting Point -130.0°C
Quantity 100mL
Chemical Name or Material n-Pentane
Show More Show Less

GHS H Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking.

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.

Danger

RUO – Research Use Only