Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Pentane, +99%, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
1L
Packaging:
AcroSeal™ Glass Bottle
109-66-0
C5H12
72.15
MFCD00009498
OFBQJSOFQDEBGM-UHFFFAOYSA-N
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
8003
CHEBI:37830
pentane
CCCCC
109-66-0
C5H12
72.15
MFCD00009498
OFBQJSOFQDEBGM-UHFFFAOYSA-N
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
8003
CHEBI:37830
pentane
CCCCC
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula CH3(CH2)3CH3
Merck Index 15, 7228
Formula Weight 72.15
Physical Form Liquid
Percent Purity ≥99%
Grade Extra Dry
Density 0.6260g/mL
Infrared Spectrum Authentic
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.358
Residue after Evaporation 0.001% max.
Solubility Solubility in water: insoluble. Other solubilities: soluble in alcohol and most other common organic, solvents
Specific Gravity 0.626
Water 0.002% max.
Flash Point −49°C
Viscosity 0.25 mPa.s (20°C)
Boiling Point 36.0°C
Color Colorless
Melting Point -130.0°C
Quantity 100mL
Chemical Name or Material n-Pentane, Extra Dry
Show More Show Less

GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking.

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.

Danger