n-Pentane, 99% min., HPLC Grade, Alfa Aesar™

Maximum level of impurities: Evaporation residue 3ppm, H2O 0.01%

Manufacturer: Alfa Aesar 22907K7

Quantity 4L

 View more versions of this product

Catalog No. AA22907K7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 109-66-0
    Physical Form Liquid
    Assay Percent Range 99% min.
    Molecular Formula CH3(CH2)3CH3
    Formula Weight 72.15
    MDL Number MFCD00009498
    Chemical Name or Material n-Pentane
    Grade HPLC Grade

    Specifications

    Boiling Point 36°C
    Physical Form Liquid
    Color Clear colorless
    Melting Point −130°C
    Formula Weight 72.15
    Quantity 4L
    Refractive Index 1.358
    Flash Point −49°C
    UN Number 1265
    DOT Information Hazard Class: 3, Packing Group: II
    Density 0.626g/cm3
    EINECS Number 203-692-4
    MDL Number MFCD00009498
    CAS 109-66-0
    Appearance Colorless liquid
    Assay Percent Range 99% min.
    Ignition Point 260°C (500°F)
    Vapor Pressure 554 hPa (416mm Hg) at 20°C
    Molecular Formula CH3(CH2)3CH3
    Health Hazard 1 H225-H304-H336-H411-H401: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Toxic to aquatic life.
    Chemical Name or Material n-Pentane
    Grade HPLC Grade

    H225-H304-H336-H411-H401: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Toxic to aquatic life.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit