Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Pentane, for analysis, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
10L
Packaging:
Aluminium Can
Glass bottle
109-66-0
C5H12
72.15
MFCD00009498
OFBQJSOFQDEBGM-UHFFFAOYSA-N
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
8003
CHEBI:37830
pentane
CCCCC
109-66-0
C5H12
72.15
MFCD00009498
OFBQJSOFQDEBGM-UHFFFAOYSA-N
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
8003
CHEBI:37830
pentane
CCCCC
For analysis
Acidity Alkalinity 0.0001 meq/g max.
Linear Formula CH3(CH2)3CH3
Merck Index 15, 7228
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and most other common organic, solvents
Calcium (Ca) 0.5ppm max.
Formula Weight 72.15
Physical Form Liquid
Percent Purity ≥99.0%
Grade Analysis
Copper (Cu) 0.05ppm max.
Density 0.6260g/mL
Iron (Fe) 0.2ppm max.
Lead (Pb) 0.05ppm max.
Magnesium (Mg) 0.1ppm max.
Packaging Glass bottle
Potassium (K) 0.2ppm max.
Refractive Index 1.358
Residue after Evaporation 0.002% max.
Silicon (Si) 0.02ppm max.
Solubility Solubility in water: insoluble. Other solubilities: soluble in alcohol and most other common organic, solvents
Sodium (Na) 1ppm max.
Specific Gravity 0.626
Total Phosphorus 0.5ppm max.
Trace Analysis 0.1ppm max.
Total Sulfur 5ppm max.
Water 0.01% max. (K.F.)
Flash Point −49°C
Viscosity 0.25 mPa.s (20°C)
Boiling Point 36.0°C
Color Undesignated
Melting Point -130.0°C
Quantity 1L
Chemical Name or Material n-Pentane
Show More Show Less

GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking.

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.

Danger