Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ n-Pentane, anhydrous, ≥99.8%, over molecular sieves, packaged under Argon in resealable ChemSeal™ bottles

Click to view available options
Quantity:
250mL
1L
109-66-0
C5H12
72.15
MFCD00009498
OFBQJSOFQDEBGM-UHFFFAOYSA-N
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
8003
CHEBI:37830
pentane
CCCCC
109-66-0
C5H12
72.15
MFCD00009498
OFBQJSOFQDEBGM-UHFFFAOYSA-N
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
8003
CHEBI:37830
pentane
CCCCC
  • Packaged under Argon
Linear Formula CH3(CH2)3CH3
UN Number UN1265
Beilstein 969132
Merck Index 14,7116
Solubility Information Miscible with water.
Formula Weight 72.15
Physical Form Liquid
Percent Purity ≥99.8%
Density 0.626 g/mL
Odor Petroleum-like
Packaging ChemSeal™ bottles
Refractive Index 1.358
Flash Point -49°C (-57°F)
Boiling Point 36°C
Color Colorless
Melting Point -130°C
Quantity 250mL
Chemical Name or Material n-Pentane, Anhydrous, over molecular sieves
Show More Show Less

GHS H Statement
H225-H304-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.

P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P331-P370+P378q-P501c

H225-H304-H336

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: PENTANES

EINECSNumber : 203-692-4

RTECSNumber : RZ9450000

TSCA : Yes

storageNote1 :

  • Packaged under Argon

Recommended Storage : Ambient temperatures