Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Pentane, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle
109-66-0
C5H12
72.151
OFBQJSOFQDEBGM-UHFFFAOYSA-N
n-pentane,pentan,skellysolve a,pentanen,pentani,amyl hydride,tetrafume,tetrakil,tetraspot,pentan polish
8003
CHEBI:37830
pentane
CCCCC
109-66-0
C5H12
72.151
OFBQJSOFQDEBGM-UHFFFAOYSA-N
n-pentane,pentan,skellysolve a,pentanen,pentani,amyl hydride,tetrafume,tetrakil,tetraspot,pentan polish
8003
CHEBI:37830
pentane
CCCCC

High quality chemicals for laboratory and specialized industrial use

Formula Weight 72.15
Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.63kg
Flash Point −40.00°C
Quantity 4L
Chemical Name or Material n-Pentane