n-Pentane, Environmental Grade, >98+%, Alfa Aesar™

Suitable For Environmental, Pesticide, and GC Analysis

Manufacturer: Alfa Aesar 40981M1

Quantity 4 x 1L

 View more versions of this product

Catalog No. AA40981M1

 N/A
    N/A

This unit of measure is invalid. 


    Chemical Identifiers

    CAS 109-66-0
    Physical Form Liquid
    Assay Percent Range >98%
    Molecular Formula CH3(CH2)3CH3
    Formula Weight 72.15
    MDL Number MFCD00009498
    Chemical Name or Material n-Pentane, Environmental Grade
    Grade Environmental Grade

    Specifications

    Physical Form Liquid
    Color Clear colorless
    Formula Weight 72.15
    Quantity 4 x 1L
    DOT Information DOT Class: 3, Packaging Group: II
    EINECS Number 203-692-4
    Ignition Point 260°C (500°F)
    Vapor Pressure 554 hPa (416mm Hg) at 20°C
    Appearance Colorless liquid
    Health Hazard 1 H225-H304-H336-H411-H401: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Toxic to aquatic life.
    Assay Percent Range >98%
    Molecular Formula CH3(CH2)3CH3
    MDL Number MFCD00009498
    CAS 109-66-0
    Chemical Name or Material n-Pentane, Environmental Grade
    Grade Environmental Grade

    H225-H304-H336-H411-H401: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Toxic to aquatic life.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit