Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Pentane, Environmental Grade, >98+%, Alfa Aesar™

Click to view available options
Quantity:
1L
4L
4 x 4L
109-66-0
C5H12
72.15
MFCD00009498
OFBQJSOFQDEBGM-UHFFFAOYSA-N
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
8003
CHEBI:37830
pentane
CCCCC
109-66-0
C5H12
72.15
MFCD00009498
OFBQJSOFQDEBGM-UHFFFAOYSA-N
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
8003
CHEBI:37830
pentane
CCCCC

Suitable For Environmental, Pesticide, and GC Analysis

Appearance Colorless liquid
Assay Percent Range >98%
Solubility Information miscible with water.
Formula Weight 72.15
Physical Form Liquid
Grade Environmental
Density 0.626
Ignition Point 260°C (500°F)
Refractive Index 1.358
Vapor Pressure 554 hPa (416mm Hg) at 20°C
Flash Point -49°C (-57°F)
Boiling Point 36°C
Color Colorless
Melting Point °C to 130°C
Quantity 1L
Chemical Name or Material n-Pentane, Environmental Grade
Show More Show Less

P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P331-P370+P378q-P501c

H225-H304-H336

DOTInformation : DOT Class: 3, Packaging Group: II

EINECSNumber : 203-692-4

RTECSNumber : RZ9450000

TSCA : Yes

Recommended Storage : Ambient temperatures