n-Pentane, Spectrophotometric Grade, >99% min., Alfa Aesar™

Manufacturer: Alfa Aesar 32449K2

Quantity 1L

 View more versions of this product

Catalog No. AA32449K2

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 109-66-0
    Physical Form Liquid
    Assay Percent Range >99%
    Molecular Formula CH3(CH2)3CH3
    Formula Weight 72.15
    MDL Number MFCD00009498
    Chemical Name or Material n-Pentane
    Grade Spectrophotometric grade

    Specifications

    Formula Weight 72.15
    Quantity 1L
    Color Colorless
    DOT Information DOT Class: 3; Packing Group: II
    EINECS Number 109-66-0
    Physical Form Liquid
    Vapor Pressure 554 hPa (416mm Hg) at 20°C
    Appearance Colorless liquid
    Assay Percent Range >99%
    Molecular Formula CH3(CH2)3CH3
    MDL Number MFCD00009498
    CAS 109-66-0
    Health Hazard 1 H225-H304-H336-H411-H401: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Toxic to aquatic life.
    Chemical Name or Material n-Pentane
    Grade Spectrophotometric grade

    H225-H304-H336-H411-H401: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Toxic to aquatic life.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit