Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ n-Pentyl 4-aminobenzoate, 98%

Click to view available options
Quantity:
5g
25g
13110-37-7
C12H17NO2
207.273
MFCD00455845
VKYWCHMXHQTCJQ-UHFFFAOYSA-N
n-amyl 4-aminobenzoate,benzoic acid, 4-amino-, pentyl ester,n-pentyl 4-aminobenzoate,4-aminobenzoic acid n-pentyl ester
25706
pentyl 4-aminobenzoate
CCCCCOC(=O)C1=CC=C(C=C1)N
13110-37-7
C12H17NO2
207.273
MFCD00455845
VKYWCHMXHQTCJQ-UHFFFAOYSA-N
n-amyl 4-aminobenzoate,benzoic acid, 4-amino-, pentyl ester,n-pentyl 4-aminobenzoate,4-aminobenzoic acid n-pentyl ester
25706
pentyl 4-aminobenzoate
CCCCCOC(=O)C1=CC=C(C=C1)N
Quantity 5g
Melting Point 51°C to 53°C
Formula Weight 207.27
Percent Purity 98%
Chemical Name or Material n-Pentyl 4-aminobenzoate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning