Alfa Aesar™ n-Pentyl benzoate, 98+%

Manufacturer: Alfa Aesar™ A10177-0E

Quantity 2500g

 View more versions of this product

Catalog No. AAA101770E

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 2049-96-9
    Molecular Formula C12H16O2
    Formula Weight 192.26
    MDL Number MFCD00048843
    Synonym n-Amyl benzoate; Benzoic acid n-pentyl ester
    Chemical Name or Material n-Pentyl benzoate

    Specifications

    Formula Weight 192.26
    MDL Number MFCD00048843
    Molecular Formula C12H16O2
    CAS 2049-96-9
    Synonym n-Amyl benzoate; Benzoic acid n-pentyl ester
    RTECS Number DH6276000
    Flash Point 130°C (266°F)
    Quantity 2500g
    TSCA TSCA
    Boiling Point 260°C
    Chemical Name or Material n-Pentyl benzoate
    Assay ≥98%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit