Alfa Aesar™ N-Phenyl-1-naphthylamine hydrobromide, 97%

Manufacturer: Alfa Aesar™ H2761506

Quantity 5g

 View more versions of this product

Catalog No. AAH2761506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 205526-65-4
    Molecular Formula C16H13N·HBr
    Formula Weight 300.2
    MDL Number MFCD00044801
    UN Number UN3077
    Synonym N-(1-Naphthyl)aniline hydrobromide
    Chemical Name or Material N-Phenyl-1-naphthylamine hydrobromide

    Specifications

    Formula Weight 300.2
    MDL Number MFCD00044801
    CAS 205526-65-4
    UN Number UN3077
    Synonym N-(1-Naphthyl)aniline hydrobromide
    Health Hazard 2 GHS H Statement
    H301-H317
    Toxic if swallowed.
    May cause an allergic skin reaction.
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Melting Point 221° to 229°C
    Quantity 5g
    Molecular Formula C16H13N·HBr
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material N-Phenyl-1-naphthylamine hydrobromide
    Assay 97%

    GHS H Statement
    H301-H317
    Toxic if swallowed.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P301+P310-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit