Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Phenyl-N-(4-piperidinyl)propanamide admixture with HCl salt, ACROS Organics™

Click to view available options
Quantity:
2.5g
Packaging:
Glass bottle
1609-66-1,22352-81-4
C14H20N2O
232.327
MFCD11106684
PMCBDBWCQQBSRJ-UHFFFAOYSA-N
norfentanyl,n-phenyl-n-piperidin-4-yl propionamide,n-phenyl-n-piperidin-4-yl-propionamide,4-n-propionylanilino piperidine,unii-2mk6d8jv6j,n-phenyl-n-4-piperidinyl propanamide,propanamide, n-phenyl-n-4-piperidinyl,n-phenyl-n-4-piperidinylpropanamide,n-phenyl-n-piperidin-4-yl propanamide,n-phenyl-n-4-piperidinylpropionamide
259381
CHEBI:62685
N-phenyl-N-piperidin-4-ylpropanamide
CCC(=O)N(C1CCNCC1)C2=CC=CC=C2
1609-66-1,22352-81-4
C14H20N2O
232.327
MFCD11106684
PMCBDBWCQQBSRJ-UHFFFAOYSA-N
norfentanyl,n-phenyl-n-piperidin-4-yl propionamide,n-phenyl-n-piperidin-4-yl-propionamide,4-n-propionylanilino piperidine,unii-2mk6d8jv6j,n-phenyl-n-4-piperidinyl propanamide,propanamide, n-phenyl-n-4-piperidinyl,n-phenyl-n-4-piperidinylpropanamide,n-phenyl-n-piperidin-4-yl propanamide,n-phenyl-n-4-piperidinylpropionamide
259381
CHEBI:62685
N-phenyl-N-piperidin-4-ylpropanamide
CCC(=O)N(C1CCNCC1)C2=CC=CC=C2
Quantity 2.5g
Color White to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Formula Weight 232.32
Physical Form Powder
Percent Purity 97%
Chemical Name or Material N-Phenyl-N-(4-piperidinyl)propanamide

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger