Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Phenyl-o-phenylenediamine 97%, ACROS Organics™

Click to view available options
Quantity:
50g
Packaging:
Glass bottle
534-85-0
C12H12N2
184.24
MFCD00007685
NFCPRRWCTNLGSN-UHFFFAOYSA-N
2-aminodiphenylamine,n-phenyl-o-phenylenediamine,n1-phenylbenzene-1,2-diamine,o-semidine,1,2-benzenediamine, n-phenyl,o-aminodiphenylamine,n-phenyl-1,2-phenylenediamine,n-phenylbenzene-1,2-diamine,o-phenylenediamine, n-phenyl,unii-g2x60k1y26
68297
2-N-phenylbenzene-1,2-diamine
C1=CC=C(C=C1)NC2=CC=CC=C2N
534-85-0
C12H12N2
184.24
MFCD00007685
NFCPRRWCTNLGSN-UHFFFAOYSA-N
2-aminodiphenylamine,n-phenyl-o-phenylenediamine,n1-phenylbenzene-1,2-diamine,o-semidine,1,2-benzenediamine, n-phenyl,o-aminodiphenylamine,n-phenyl-1,2-phenylenediamine,n-phenylbenzene-1,2-diamine,o-phenylenediamine, n-phenyl,unii-g2x60k1y26
68297
2-N-phenylbenzene-1,2-diamine
C1=CC=C(C=C1)NC2=CC=CC=C2N
Melting Point 77°C to 82°C
Color Brown-Gray or Pink-Violet
Quantity 50g
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C6H5NHC6H4NH2
Beilstein 13, 16
Formula Weight 184.24
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material N-Phenyl-o-phenylenediamine, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.