Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Phenyl-p-phenylenediamine, 98%

Click to view available options
Quantity:
100g
250g
1000g
101-54-2
C12H12N2
184.242
MFCD00007850
ATGUVEKSASEFFO-UHFFFAOYSA-N
4-aminodiphenylamine,n-phenyl-p-phenylenediamine,n1-phenylbenzene-1,4-diamine,p-anilinoaniline,azosalt r,p-semidine,p-aminodiphenylamine,luxan black r,semidine,n-phenyl-1,4-phenylenediamine
7564
CHEBI:59038
4-N-phenylbenzene-1,4-diamine
C1=CC=C(C=C1)NC2=CC=C(C=C2)N
101-54-2
C12H12N2
184.242
MFCD00007850
ATGUVEKSASEFFO-UHFFFAOYSA-N
4-aminodiphenylamine,n-phenyl-p-phenylenediamine,n1-phenylbenzene-1,4-diamine,p-anilinoaniline,azosalt r,p-semidine,p-aminodiphenylamine,luxan black r,semidine,n-phenyl-1,4-phenylenediamine
7564
CHEBI:59038
4-N-phenylbenzene-1,4-diamine
C1=CC=C(C=C1)NC2=CC=C(C=C2)N
Applications
p-Anilinoaniline is an antioxidant that has shown to cause a reduction of NOx emissions from soybean biodiesel powered DI diesel engines but has increased CO and HC emissions. It is also used to produce dyes, pharmaceuticals, and photographic chemicals and frmerly used as a hair dye.

Solubility
Soluble in water at 20°C 0.6g/L. Soluble in (10 mg/mL) ethanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Melting Point 72°C to 76°C
Boiling Point 354°C
Flash Point 193°C (379°F)
Quantity 100g
UN Number UN3077
Beilstein 908935
Formula Weight 184.24
Percent Purity 98%
Chemical Name or Material N-Phenyl-p-phenylenediamine

GHS H Statement
H301-H315-H319-H317-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : ST3150000

TSCA : TSCA