Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Phenylbis(trifluoromethanesulfonimide), 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
37595-74-7
C8H5F6NO4S2
357.25
MFCD00000404
DIOHEXPTUTVCNX-UHFFFAOYSA-N
n,n-bis trifluoromethylsulfonyl aniline,phenyl triflimide,n-phenyl-bis trifluoromethanesulfonimide,n-phenyltrifluoromethanesulfonimide,n,n-bis-trifluoromethanesulfonyl aniline,1,1,1-trifluoro-n-phenyl-n-trifluoromethylsulfonyl methanesulfonamide,n-phenylbis trifluoromethanesulfonimide,n-phenylbis trifluoromethanesulphonimide,1,1,1-trifluoro-n-phenyl-n-trifluoromethyl sulfonyl methanesulfonamide,n,n-bis trifluoromethanesulfonyl aniline
142176
1,1,1-trifluoro-N-phenyl-N-(trifluoromethylsulfonyl)methanesulfonamide
C1=CC=C(C=C1)N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F
37595-74-7
C8H5F6NO4S2
357.25
MFCD00000404
DIOHEXPTUTVCNX-UHFFFAOYSA-N
n,n-bis trifluoromethylsulfonyl aniline,phenyl triflimide,n-phenyl-bis trifluoromethanesulfonimide,n-phenyltrifluoromethanesulfonimide,n,n-bis-trifluoromethanesulfonyl aniline,1,1,1-trifluoro-n-phenyl-n-trifluoromethylsulfonyl methanesulfonamide,n-phenylbis trifluoromethanesulfonimide,n-phenylbis trifluoromethanesulphonimide,1,1,1-trifluoro-n-phenyl-n-trifluoromethyl sulfonyl methanesulfonamide,n,n-bis trifluoromethanesulfonyl aniline
142176
1,1,1-trifluoro-N-phenyl-N-(trifluoromethylsulfonyl)methanesulfonamide
C1=CC=C(C=C1)N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F
Coupling reagent
Packaging Glass bottle
Quantity 25g
Infrared Spectrum Authentic
Melting Point 99.0°C to 105.0°C
Color White or Colorless
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C6H5N(SO2CF3)2
Merck Index 15,7427
Formula Weight 357.25
Physical Form Crystals or Crystalline Powder
Percent Purity 97%
Chemical Name or Material N-Phenylbis(trifluoromethanesulfonimide)
Show More Show Less

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning