Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Phenylcarbazole, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
1150-62-5
C18H13N
243.31
VIJYEGDOKCKUOL-UHFFFAOYSA-N
n-phenylcarbazole,9-phenyl-9h-carbazole,9h-carbazole, 9-phenyl,carbazole, 9-phenyl,carbazol, 9-phenyl,9-phenyl carbazole,pubchem23849,9h-carbazole,9-phenyl,9-phenylcarbazole,acmc-2099pd
70851
9-phenylcarbazole
C1=CC=C(C=C1)N2C3=CC=CC=C3C4=CC=CC=C42
1150-62-5
C18H13N
243.31
VIJYEGDOKCKUOL-UHFFFAOYSA-N
n-phenylcarbazole,9-phenyl-9h-carbazole,9h-carbazole, 9-phenyl,carbazole, 9-phenyl,carbazol, 9-phenyl,9-phenyl carbazole,pubchem23849,9h-carbazole,9-phenyl,9-phenylcarbazole,acmc-2099pd
70851
9-phenylcarbazole
C1=CC=C(C=C1)N2C3=CC=CC=C3C4=CC=CC=C42
Melting Point 94.0°C to 97.0°C
Quantity 100g
Assay Percent Range 96% min. (GC)
Beilstein 20,I,164
Formula Weight 243.31
Percent Purity 97%
Chemical Name or Material N-Phenylcarbazole

GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTRE or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Danger

EINECSNumber : 214-564-2