Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Phenyldiethanolamine, 95%, ACROS Organics™

Click to view available options
Quantity:
500g
1kg
2.5kg
Packaging:
Glass bottle
120-07-0,62-53-3
C10H15NO2
181.23
MFCD00002845
OJPDDQSCZGTACX-UHFFFAOYSA-N
n-phenyldiethanolamine,2,2'-phenylimino diethanol,phenyldiethanolamine,diethanolaniline,diethanolphenylamine,diethanolaminobenzene,phenyl diethanolamine,n,n-diethanolaniline,n,n-dioxyethylaniline,n,n-bis 2-hydroxyethyl aniline
8416
2-[N-(2-hydroxyethyl)anilino]ethanol
C1=CC=C(C=C1)N(CCO)CCO
120-07-0,62-53-3
C10H15NO2
181.23
MFCD00002845
OJPDDQSCZGTACX-UHFFFAOYSA-N
n-phenyldiethanolamine,2,2'-phenylimino diethanol,phenyldiethanolamine,diethanolaniline,diethanolphenylamine,diethanolaminobenzene,phenyl diethanolamine,n,n-diethanolaniline,n,n-dioxyethylaniline,n,n-bis 2-hydroxyethyl aniline
8416
2-[N-(2-hydroxyethyl)anilino]ethanol
C1=CC=C(C=C1)N(CCO)CCO
Boiling Point 270°C
Flash Point 200°C
Viscosity 1.27 mPa.s (200°C)
Infrared Spectrum Authentic
Quantity 1kg
Melting Point 55°C to 59°C
Color White to Brown
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Linear Formula (HOCH2CH2)2NC6H5
Solubility Information Solubility in water: 45g/L (20°C). Other solubilities: freely soluble in ethanol,acetone,ethyl acetate,and ether (20°C)
Formula Weight 181.23
Physical Form Crystalline Mass, Crystals or Chunks
Percent Purity 95%
Chemical Name or Material N-Phenyldiethanolamine, 90-95%
Show More Show Less

GHS H Statement:
Harmful to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye damage.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 204-368-5

RTECSNumber : KM2100000

TSCA : TSCA

Recommended Storage : This product tends to darken during storage