Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Phenyldiethanolamine, 95%, Thermo Scientific™

Catalog Number p-181837
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
120-07-0, 62-53-3
C10H15NO2
181.24
MFCD00002845
OJPDDQSCZGTACX-UHFFFAOYSA-N
n-phenyldiethanolamine, 2,2'-phenylimino diethanol, phenyldiethanolamine, diethanolaniline, diethanolphenylamine, diethanolaminobenzene, phenyl diethanolamine, n,n-diethanolaniline, n,n-dioxyethylaniline, n,n-bis 2-hydroxyethyl aniline
8416
2-[N-(2-hydroxyethyl)anilino]ethanol
OCCN(CCO)C1=CC=CC=C1
120-07-0, 62-53-3
C10H15NO2
181.24
MFCD00002845
OJPDDQSCZGTACX-UHFFFAOYSA-N
n-phenyldiethanolamine, 2,2'-phenylimino diethanol, phenyldiethanolamine, diethanolaniline, diethanolphenylamine, diethanolaminobenzene, phenyl diethanolamine, n,n-diethanolaniline, n,n-dioxyethylaniline, n,n-bis 2-hydroxyethyl aniline
8416
2-[N-(2-hydroxyethyl)anilino]ethanol
OCCN(CCO)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 55°C to 59°C
Color White to Brown
Boiling Point 270°C
Flash Point 200°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (HOCH2CH2)2NC6H5
Quantity 500 g
Solubility Information Solubility in water: 45g/L (20°C). Other solubilities: freely soluble in ethanol,acetone,ethyl acetate,and ether (20°C)
Viscosity 1.27 mPa.s (200°C)
Formula Weight 181.23
Percent Purity 95%
Physical Form Crystalline Mass, Crystals or Chunks
Chemical Name or Material N-Phenyldiethanolamine, 90-95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-368-5
RTECSNumber KM2100000
TSCA TSCA
Recommended Storage This product tends to darken during storage

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit