Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Phenylethanolamine, 98%

Click to view available options
Quantity:
500g
2500g
122-98-5
C8H11NO
137.182
MFCD00002832
MWGATWIBSKHFMR-UHFFFAOYSA-N
2-phenylamino ethanol,n-phenylethanolamine,n-2-hydroxyethyl aniline,ethanol, 2-phenylamino,2-phenylamino ethan-1-ol,ethanol, 2-anilino,beta-anilinoethanol,aniline, n-2-hydroxyethyl,n-2-hydroxyethyl phenylamine,emery 5700
31235
2-anilinoethanol
C1=CC=C(C=C1)NCCO
122-98-5
C8H11NO
137.182
MFCD00002832
MWGATWIBSKHFMR-UHFFFAOYSA-N
2-phenylamino ethanol,n-phenylethanolamine,n-2-hydroxyethyl aniline,ethanol, 2-phenylamino,2-phenylamino ethan-1-ol,ethanol, 2-anilino,beta-anilinoethanol,aniline, n-2-hydroxyethyl,n-2-hydroxyethyl phenylamine,emery 5700
31235
2-anilinoethanol
C1=CC=C(C=C1)NCCO
Applications
N-Phenylethanolamine is used as an intermediate for dyes and other organic compounds. Further, it is used as a substrate for human olfactory uridine 5′-diphospho-glucuronosyltransferase.

Solubility
Slightly miscible with water.

Notes
Incompatible with acids, acid anhydrides, acid chlorides and chloroformates.
Quantity 2500g
Boiling Point 284°C to 286°C
pH 10.2
Flash Point 153°C (307°F)
Odor Amine-like
Melting Point -30°C
UN Number UN2810
Beilstein 774672
Formula Weight 137.18
Percent Purity 98%
Chemical Name or Material N-Phenylethanolamine
Show More Show Less

GHS H Statement
H310-H351-H373-H319-H317
Fatal in contact with skin.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P260-P305+P351+P338-P361-P302+P350-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

RTECSNumber : KJ7175000

TSCA : TSCA