Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-Phenylethanolamine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2500 g
Description
N-Phenylethanolamine is used as an intermediate for dyes and other organic compounds. Further, it is used as a substrate for human olfactory uridine 5'-diphospho-glucuronosyltransferase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Phenylethanolamine is used as an intermediate for dyes and other organic compounds. Further, it is used as a substrate for human olfactory uridine 5′-diphospho-glucuronosyltransferase.
Solubility
Slightly miscible with water.
Notes
Incompatible with acids, acid anhydrides, acid chlorides and chloroformates.
N-Phenylethanolamine is used as an intermediate for dyes and other organic compounds. Further, it is used as a substrate for human olfactory uridine 5′-diphospho-glucuronosyltransferase.
Solubility
Slightly miscible with water.
Notes
Incompatible with acids, acid anhydrides, acid chlorides and chloroformates.
Specifications
Melting Point | -30°C |
Density | 1.087 |
pH | 10.2 |
Boiling Point | 284°C to 286°C |
Flash Point | 153°C (307°F) |
Odor | Amine-like |
Refractive Index | 1.579 |
Quantity | 500 g |
UN Number | UN2810 |
Beilstein | 774672 |
Show More |
Safety and Handling
Hazard Category | H311-H319 |
Hazard Statement | GHS H Statement H310-H351-H373-H319-H317 Fatal in contact with skin. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure. Causes serious eye irritation. May cause an allergic skin reaction. |
Precautionary Statement | P264b-P280-P302+P352-P305+P351+P338-P312-P361-P363-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S. |
EINECSNumber | 204-588-1 |
RTECSNumber | KJ7175000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction