Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Phenylethylenediamine, 99%, ACROS Organics™

Click to view available options
Quantity:
10mL
50mL
Packaging:
Glass bottle
1664-40-0
C8H12N2
136.2
MFCD00008162
OCIDXARMXNJACB-UHFFFAOYSA-N
n-phenylethylenediamine,n1-phenylethane-1,2-diamine,n-2-aminoethyl aniline,1,2-ethanediamine, n-phenyl,ethylenediamine, n-phenyl,n-phenyl-1,2-ethanediamine,benzenamine, n-2-aminoethyl,n-phenylethane-1,2-diamine,1,2-ethanediamine, n1-phenyl,2-aminoethyl phenylamine
74270
N'-phenylethane-1,2-diamine
C1=CC=C(C=C1)NCCN
1664-40-0
C8H12N2
136.2
MFCD00008162
OCIDXARMXNJACB-UHFFFAOYSA-N
n-phenylethylenediamine,n1-phenylethane-1,2-diamine,n-2-aminoethyl aniline,1,2-ethanediamine, n-phenyl,ethylenediamine, n-phenyl,n-phenyl-1,2-ethanediamine,benzenamine, n-2-aminoethyl,n-phenylethane-1,2-diamine,1,2-ethanediamine, n1-phenyl,2-aminoethyl phenylamine
74270
N'-phenylethane-1,2-diamine
C1=CC=C(C=C1)NCCN
Quantity 50mL
Flash Point >112°C
Infrared Spectrum Authentic
Color Yellow
Boiling Point 262.0°C to 264.0°C
Density 1.0400g/mL
Packaging Glass bottle
Refractive Index 1.5855 to 1.5875
Specific Gravity 1.04
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C6H5NHCH2CH2NH2
Beilstein 12, 543
Formula Weight 136.2
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material N-Phenylethylenediamine
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger