Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Phenylglycine ethyl ester, 99%

Click to view available options
Quantity:
10g
50g
250g
2216-92-4
C10H13NO2
179.219
MFCD00038315
MLSGRWDEDYJNER-UHFFFAOYSA-N
n-phenylglycine ethyl ester,ethyl phenylglycinate,ethyl 2-phenylamino acetate,ethyl n-phenylglycinate,glycine, n-phenyl-, ethyl ester,unii-7xl9t8bk0u,ethyl anilinoacetate,n-phenylglycineethylester,n-phenylglycine ethyl,7xl9t8bk0u
75190
ethyl 2-anilinoacetate
CCOC(=O)CNC1=CC=CC=C1
2216-92-4
C10H13NO2
179.219
MFCD00038315
MLSGRWDEDYJNER-UHFFFAOYSA-N
n-phenylglycine ethyl ester,ethyl phenylglycinate,ethyl 2-phenylamino acetate,ethyl n-phenylglycinate,glycine, n-phenyl-, ethyl ester,unii-7xl9t8bk0u,ethyl anilinoacetate,n-phenylglycineethylester,n-phenylglycine ethyl,7xl9t8bk0u
75190
ethyl 2-anilinoacetate
CCOC(=O)CNC1=CC=CC=C1
Merck Index 14,7292
Formula Weight 179.22
Percent Purity 99%
Melting Point 55°C to 58°C
Quantity 250g
Chemical Name or Material N-Phenylglycine ethyl ester

TSCA : TSCA

Recommended Storage : 2° to 8°C