Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Phthaloyl-L-glycine N-succinimidyl ester, 95%

Click to view available options
Quantity:
1g
5g
3397-29-3
C14H10N2O6
302.242
MFCD00190994
NQZZTHXHURDXOR-UHFFFAOYSA-N
pht-gly-osu,2,5-dioxopyrrolidin-1-yl 2-1,3-dioxoisoindol-2-yl acetate,2,5-dioxoazolidinyl 2-1,3-dioxobenzo c azolidin-2-yl acetate,1,3-dioxo-2h-isoindole-2-acetic acid 2,5-dioxopyrrolidine-1-yl ester,2,5-dioxopyrrolidin-1-yl 2-1,3-dioxoisoindol-2-yl acetate,2h-isoindole-2-acetic acid, 1,3-dihydro-1,3-dioxo-, 2,5-dioxo-1-pyrrolidinyl ester
2063302
(2,5-dioxopyrrolidin-1-yl) 2-(1,3-dioxoisoindol-2-yl)acetate
C1CC(=O)N(C1=O)OC(=O)CN2C(=O)C3=CC=CC=C3C2=O
3397-29-3
C14H10N2O6
302.242
MFCD00190994
NQZZTHXHURDXOR-UHFFFAOYSA-N
pht-gly-osu,2,5-dioxopyrrolidin-1-yl 2-1,3-dioxoisoindol-2-yl acetate,2,5-dioxoazolidinyl 2-1,3-dioxobenzo c azolidin-2-yl acetate,1,3-dioxo-2h-isoindole-2-acetic acid 2,5-dioxopyrrolidine-1-yl ester,2,5-dioxopyrrolidin-1-yl 2-1,3-dioxoisoindol-2-yl acetate,2h-isoindole-2-acetic acid, 1,3-dihydro-1,3-dioxo-, 2,5-dioxo-1-pyrrolidinyl ester
2063302
(2,5-dioxopyrrolidin-1-yl) 2-(1,3-dioxoisoindol-2-yl)acetate
C1CC(=O)N(C1=O)OC(=O)CN2C(=O)C3=CC=CC=C3C2=O
Beilstein 1506140
Formula Weight 302.24
Physical Form Solid
Percent Purity 95%
Quantity 1g
Sensitivity Moisture Sensitive
Chemical Name or Material N-Phthaloyl-L-glycine N-succinimidyl ester