Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Phthaloyl-L-phenylalanine, 98%

Click to view available options
Quantity:
5g
25g
5123-55-7
C17H13NO4
295.294
MFCD00069738
VAYRSTHMTWUHGE-AWEZNQCLSA-N
n-phthaloyl-l-phenylalanine,n-pht-phe-oh,phthaloyl-l-phenylalanine,n-phthalyl-l-phenylalanine,s-n-phthaloylphenylalanine,n-phthaloyl-phe-oh,2s-2-1,3-dioxoisoindol-2-yl-3-phenylpropanoic acid,2s-2-1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl-3-phenylpropanoic acid,pht-phe-oh,ncistruc1_000419
334207
(2S)-2-(1,3-dioxoisoindol-2-yl)-3-phenylpropanoic acid
C1=CC=C(C=C1)CC(C(=O)O)N2C(=O)C3=CC=CC=C3C2=O
5123-55-7
C17H13NO4
295.294
MFCD00069738
VAYRSTHMTWUHGE-AWEZNQCLSA-N
n-phthaloyl-l-phenylalanine,n-pht-phe-oh,phthaloyl-l-phenylalanine,n-phthalyl-l-phenylalanine,s-n-phthaloylphenylalanine,n-phthaloyl-phe-oh,2s-2-1,3-dioxoisoindol-2-yl-3-phenylpropanoic acid,2s-2-1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl-3-phenylpropanoic acid,pht-phe-oh,ncistruc1_000419
334207
(2S)-2-(1,3-dioxoisoindol-2-yl)-3-phenylpropanoic acid
C1=CC=C(C=C1)CC(C(=O)O)N2C(=O)C3=CC=CC=C3C2=O
Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in methanol. (almost transparency)

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Beilstein 90080
Formula Weight 295.29
Physical Form Powder
Percent Purity 98%
Color White
Melting Point 181°C to 185°C
Quantity 25g
Sensitivity Hygroscopic
Chemical Name or Material N-Phthaloyl-L-phenylalanine

storageNote1 : 2° to 8°C