Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Piperidineethanol 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
3040-44-6
C7H15NO
129.2
MFCD00006512
KZTWONRVIPPDKH-UHFFFAOYSA-N
2-piperidinoethanol,1-piperidineethanol,2-piperidin-1-yl ethanol,1-2-hydroxyethyl piperidine,n-2-hydroxyethyl piperidine,n-piperidineethanol,2-1-piperidinyl ethanol,n-hydroxyethyl piperidine,beta-piperidylethanol,beta-piperidinoethanol
18232
CHEBI:61238
2-piperidin-1-ylethanol
C1CCN(CC1)CCO
3040-44-6
C7H15NO
129.2
MFCD00006512
KZTWONRVIPPDKH-UHFFFAOYSA-N
2-piperidinoethanol,1-piperidineethanol,2-piperidin-1-yl ethanol,1-2-hydroxyethyl piperidine,n-2-hydroxyethyl piperidine,n-piperidineethanol,2-1-piperidinyl ethanol,n-hydroxyethyl piperidine,beta-piperidylethanol,beta-piperidinoethanol
18232
CHEBI:61238
2-piperidin-1-ylethanol
C1CCN(CC1)CCO
Melting Point 16.7°C
Boiling Point 198°C to 203°C
Flash Point 77°C
Viscosity 20.9 mPa.s (20°C)
Specific Gravity 0.973
Density 0.9730g/mL
Refractive Index 1.4794 to 1.4814
Infrared Spectrum Authentic
Color Brown
Quantity 100g
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 20, 25
Solubility Information Solubility in water: miscible
Formula Weight 129.2
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material N-Piperidineethanol, 99%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 221-244-6

RTECSNumber : TM8052000

TSCA : TSCA